Assay Details
Gene Symbol: Tatdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175151.4 | NP_780360.1 | 7 - 8 | 497 | 101 | |
GenBank mRNA | AK010232.1 | - | 6 - 7 | 582 | 101 | |
AK042248.1 | - | 7 - 8 | 498 | 101 | ||
AK161972.1 | - | 7 - 8 | 582 | 101 | ||
BC061187.1 | - | 7 - 8 | 494 | 30277245 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
