Assay Details
| Gene Symbol: Wisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018865.2 | NP_061353.1 | 3 - 4 | 785 | 86 | |
| GenBank mRNA | AB004873.1 | - | 3 - 4 | 785 | 86 | |
| AF100777.1 | - | 3 - 4 | 690 | 86 | ||
| AK013300.1 | - | 3 - 4 | 809 | 86 | ||
| AK148457.1 | - | 3 - 4 | 807 | 86 | ||
| BC048791.1 | - | 3 - 4 | 599 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wisp1
Genomic Map