Assay Details
| Gene Symbol: Cdc37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016651.1 | - | 6 - 7 | 1104 | 103 | |
| AK038578.1 | - | 6 - 7 | 926 | 103 | ||
| AK079556.1 | - | 6 - 7 | 925 | 103 | ||
| AK165398.1 | - | 6 - 7 | 1090 | 103 | ||
More Information
Set Membership: |
|

Cdc37l1
Genomic Map