Assay Details
Gene Symbol: Ptov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133949.1 | NP_598710.1 | 8 - 9 | 1145 | 80 | |
GenBank mRNA | AK178502.1 | - | 144 | 80 | ||
BC008543.1 | - | 8 - 9 | 1021 | 3496037 | 80 | |
BC024632.1 | - | 8 - 9 | 1145 | 4222241 | 80 | |
BC062108.1 | - | 2 - 3 | 1544 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
