Assay Details
Gene Symbol: Ptov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133949.1 | NP_598710.1 | 9 - 10 | 1203 | 68 | |
GenBank mRNA | BC008543.1 | - | 9 - 10 | 1079 | 3496037 | 68 |
BC024632.1 | - | 9 - 10 | 1203 | 4222241 | 68 | |
BC062108.1 | - | 3 - 4 | 1602 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
