Assay Details
Gene Symbol: Prrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022999.1 | NP_075375.1 | 1 - 2 | 64 | 91 | |
GenBank mRNA | AK005453.1 | - | 2 - 3 | 108 | 91 | |
BC025098.1 | - | 1 - 2 | 64 | 4159863 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC025098.1, NM_022999.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
