Assay Details
| Gene Symbol: Prrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022999.1 | NP_075375.1 | 6 - 7 | 704 | 96 | |
| GenBank mRNA | AF220205.1 | - | 2 - 3 | 233 | 96 | |
| AK005453.1 | - | 7 - 8 | 747 | 96 | ||
| BC025098.1 | - | 6 - 7 | 704 | 4159863 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090921 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090924 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090912 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090923 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090920 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177112 | BC025098.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177112 | NM_022999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177112 | AK005453.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | BC025098.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | NM_022999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | AK005453.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | AK011086.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | BC025098.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | NM_022999.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | AK005453.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P329917 | AK011086.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561774 | AK013653.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14535 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173822 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177112 | BC025098.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177112 | NM_022999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177112 | AK005453.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | BC025098.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | NM_022999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | AK005453.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | AK011086.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | BC025098.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | NM_022999.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | AK005453.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P329917 | AK011086.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284233 | BC025098.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284233 | NM_022999.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284233 | AK005453.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P284233 | AK011086.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090955 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06181812 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173822 | - | - |
| Whole Mouse Genome G4122A | A_52_P329917 | BC025098.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | NM_022999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | AK005453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | AK011086.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | BC025098.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | NM_022999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | AK005453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P329917 | AK011086.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284233 | BC025098.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284233 | NM_022999.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284233 | AK005453.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P284233 | AK011086.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113173 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05892822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05892829 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06638050 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05892834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090912 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090921 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090972 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090923 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090924 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25090915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06638046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06541577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06541569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05892834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06032097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06541580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06232153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06541579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06232150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06450781 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563070 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424329_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432763_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563068 | - | - |
| Mouse Genome 430 2.0 Array | 1424329_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432763_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432763_at | - | - |
| Mouse Genome 430 2.0 Array | 1424329_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65116_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 65116_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700026804.mm.1 | NM_022999.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026804.mm.1 | NM_022999.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026804.mm.1 | TC0700002987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026807.mm.1 | NM_022999.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026807.mm.1 | NM_022999.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026807.mm.1 | TC0700002987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026808.mm.1 | NM_022999.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026808.mm.1 | NM_022999.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026808.mm.1 | TC0700002987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026806.mm.1 | NM_022999.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026806.mm.1 | NM_022999.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026806.mm.1 | TC0700002987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700026802.mm.1 | NM_022999.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026802.mm.1 | NM_022999.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700026802.mm.1 | TC0700002987.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424329_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424329_a_at | - | - |
| Murine Genome U74v2 Set | 94924_PM_at | - | - |
| Murine Genome U74v2 Set | 162142_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162142_f_at | - | - |
| Murine Genome U74v2 Set | 94924_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424329_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424329_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432763_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424329_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424329_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432763_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 303758 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4540095 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004540095 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245984 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245984 | - | - |
| mouse-6 v1.1 (Illumina) | scl31397.7.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4540095 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31397.7.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540095 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245984 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245984 | - | - |

Prrg2
Genomic Map