Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005045.2 | - | 3 - 4 | 400 | 64 | |
| AK011564.1 | - | 3 - 4 | 356 | 64 | ||
| AK014311.2 | - | 3 - 4 | 397 | 64 | ||
| AK077425.1 | - | 3 - 4 | 402 | 64 | ||
| AK133115.1 | - | 3 - 4 | 401 | 64 | ||
| AK146729.1 | - | 3 - 4 | 401 | 64 | ||
| AK148013.1 | - | 3 - 4 | 427 | 64 | ||
| BC049128.1 | - | 3 - 4 | 366 | 64 | ||
| BC051414.1 | - | 2 - 3 | 201 | 64 | ||
| BC131994.1 | - | 3 - 4 | 293 | 40130271 | 64 | |
| BC132020.1 | - | 3 - 4 | 293 | 40130297 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snrnp70
Genomic Map