Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005045.2 | - | 4 - 5 | 455 | 63 | |
| AK011564.1 | - | 4 - 5 | 411 | 63 | ||
| AK014311.2 | - | 4 - 5 | 452 | 63 | ||
| AK077425.1 | - | 4 - 5 | 457 | 63 | ||
| AK133115.1 | - | 4 - 5 | 456 | 63 | ||
| AK146729.1 | - | 4 - 5 | 456 | 63 | ||
| AK148013.1 | - | 4 - 5 | 482 | 63 | ||
| BC049128.1 | - | 4 - 5 | 421 | 63 | ||
| BC051414.1 | - | 3 - 4 | 256 | 63 | ||
| BC131994.1 | - | 4 - 5 | 348 | 40130271 | 63 | |
| BC132020.1 | - | 4 - 5 | 348 | 40130297 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snrnp70
Genomic Map