Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005045.2 | - | 6 - 7 | 591 | 72 | |
| AK011564.1 | - | 6 - 7 | 547 | 72 | ||
| AK012272.1 | - | 4 - 5 | 240 | 72 | ||
| AK014311.2 | - | 6 - 7 | 588 | 72 | ||
| AK077425.1 | - | 6 - 7 | 593 | 72 | ||
| AK133115.1 | - | 6 - 7 | 592 | 72 | ||
| AK146729.1 | - | 6 - 7 | 592 | 72 | ||
| AK148013.1 | - | 6 - 7 | 618 | 72 | ||
| BC049128.1 | - | 6 - 7 | 557 | 72 | ||
| BC051414.1 | - | 5 - 6 | 392 | 72 | ||
| BC131994.1 | - | 6 - 7 | 484 | 40130271 | 72 | |
| BC132020.1 | - | 6 - 7 | 484 | 40130297 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snrnp70
Genomic Map