Assay Details
| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013520.3 | NP_038548.3 | 1 - 2 | 55 | 53 | |
| NR_102725.1 | 1 - 2 | 55 | 53 | |||
| GenBank mRNA | AK020105.1 | - | 1 - 2 | 55 | 53 | |
| BC019801.1 | - | 1 - 2 | 40 | 5132319 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020105.1, BC019801.1, NM_013520.3 transcripts.
Set Membership: |
|

Flt3l
Genomic Map