Assay Details
| Gene Symbol: Paqr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198422.2 | NP_940814.1 | 5 - 6 | 948 | 84 | |
| GenBank mRNA | AK149213.1 | - | 5 - 6 | 927 | 84 | |
| AK172689.1 | - | 5 - 6 | 904 | 84 | ||
| AY424292.1 | - | 5 - 6 | 948 | 84 | ||
| BC120853.1 | - | 5 - 6 | 933 | 40129776 | 84 | |
| BC120855.1 | - | 5 - 6 | 934 | 40129778 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paqr3
Genomic Map