Assay Details
| Gene Symbol: Ldha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136069.2 | NP_001129541.2 | 3 - 4 | 577 | 82 | |
| NM_010699.2 | NP_034829.1 | 3 - 4 | 384 | 82 | ||
| NR_102727.1 | 3 - 4 | 384 | 82 | |||
| GenBank mRNA | AK089929.1 | - | 3 - 4 | 348 | 82 | |
| AK146121.1 | - | 3 - 4 | 345 | 82 | ||
| AK149919.1 | - | 3 - 4 | 345 | 82 | ||
| AK150156.1 | - | 3 - 4 | 348 | 82 | ||
| AK167789.1 | - | 3 - 4 | 346 | 82 | ||
| AK167944.1 | - | 3 - 4 | 346 | 82 | ||
| AK168346.1 | - | 3 - 4 | 345 | 82 | ||
| AK168920.1 | - | 3 - 4 | 350 | 82 | ||
| AK170386.1 | - | 3 - 4 | 349 | 82 | ||
| AK170708.1 | - | 3 - 4 | 346 | 82 | ||
| AK171373.1 | - | 3 - 4 | 347 | 82 | ||
| AK182326.1 | - | 1 - 2 | 91 | 82 | ||
| AK189943.1 | - | 1 - 2 | 91 | 82 | ||
| AK194938.1 | - | 1 - 2 | 91 | 82 | ||
| AK200450.1 | - | 1 - 2 | 91 | 82 | ||
| AK201084.1 | - | 1 - 2 | 91 | 82 | ||
| AK203794.1 | - | 1 - 2 | 82 | |||
| AK204450.1 | - | 1 - 2 | 91 | 82 | ||
| AK206570.1 | - | 1 - 2 | 91 | 82 | ||
| BC066858.1 | - | 3 - 4 | 331 | 82 | ||
| BC094019.1 | - | 3 - 4 | 301 | 4195220 | 82 | |
| BC094428.1 | - | 3 - 4 | 338 | 30700835 | 82 | |
| U13687.1 | - | 3 - 4 | 444 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ldha
Genomic Map