Assay Details
| Gene Symbol: Suds3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122666.2 | NP_001116138.1 | 2 - 3 | 384 | 72 | |
| NM_178622.5 | NP_848737.3 | 2 - 3 | 384 | 72 | ||
| GenBank mRNA | AF469109.1 | - | 2 - 3 | 324 | 72 | |
| AK010274.1 | - | 2 - 3 | 287 | 72 | ||
| AK043652.1 | - | 2 - 3 | 327 | 72 | ||
| AK045475.1 | - | 2 - 3 | 338 | 72 | ||
| AK138947.1 | - | 2 - 3 | 335 | 72 | ||
| AK150633.1 | - | 2 - 3 | 322 | 72 | ||
| BC055764.1 | - | 2 - 3 | 337 | 6402735 | 72 | |
| BC062176.1 | - | 2 - 3 | 251 | 6514171 | 72 | |
| CJ094124.1 | - | 2 - 3 | 390 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Suds3
Genomic Map