Assay Details
Gene Symbol: Zfp707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081065.1 | NP_001074534.1 | 2 - 3 | 305 | 81 | |
GenBank mRNA | AK137962.1 | - | 2 - 3 | 374 | 81 | |
AK153958.1 | - | 2 - 3 | 255 | 81 | ||
BC026404.1 | - | 2 - 3 | 271 | 4210825 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
