Assay Details
| Gene Symbol: Acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007383.3 | NP_031409.2 | 1 - 2 | 120 | 92 | |
| GenBank mRNA | AK155361.1 | - | 1 - 2 | 99 | 92 | |
| BC016259.1 | - | 1 - 2 | 87 | 4913813 | 92 | |
| BY077302.1 | - | 1 - 2 | 121 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155361.1, BC016259.1, NM_007383.3 transcripts.
Set Membership: |
|

Acads
Genomic Map