Assay Details
| Gene Symbol: Gm38413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044041.1 | - | 8 - 9 | 1472 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A930011G23 gene | A930011G23Rik | -5.9kb | Sense | Chr. 5 - 99297244 - 99729060 on NCBI Build GRCm38 | |
| Downstream | heterogeneous nuclear ribonucleoprotein D | Hnrnpd |
Auf1, Hnrpd |
219.9kb | Sense | Chr. 5 - 99955935 - 99979015 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm38413
Genomic Map