Assay Details
| Gene Symbol: Gstm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160411.1 | NP_001153883.1 | 4 - 5 | 340 | 62 | |
| NM_026764.3 | NP_081040.1 | 5 - 6 | 545 | 62 | ||
| GenBank mRNA | AF464943.1 | - | 5 - 6 | 375 | 62 | |
| AF501320.1 | - | 5 - 6 | 361 | 62 | ||
| AK003418.1 | - | 4 - 5 | 341 | 62 | ||
| BC030444.1 | - | 5 - 6 | 492 | 5373062 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gstm4
Genomic Map