Assay Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029012.2 | NP_083288.2 | 9 - 10 | 1041 | 66 | |
| GenBank mRNA | AK014709.1 | - | 9 - 10 | 1379 | 66 | |
| BC023131.1 | - | 5 - 6 | 656 | 66 | ||
| BC086646.1 | - | 9 - 10 | 1406 | 6817386 | 66 | |
| CD806412.1 | - | 5 - 6 | 658 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sppl3
Genomic Map