Assay Details
Gene Symbol: Zfp760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008501.2 | NP_001008501.2 | 1 - 2 | 157 | 75 | |
GenBank mRNA | BC029103.1 | - | 1 - 2 | 163 | 75 | |
BC085182.1 | - | 1 - 2 | 157 | 30653533 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC029103.1, BC085182.1, NM_001008501.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
