Assay Details
Gene Symbol: Zfp760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008501.2 | NP_001008501.2 | 3 - 4 | 355 | 85 | |
GenBank mRNA | BC085182.1 | - | 3 - 4 | 355 | 30653533 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085182.1, NM_001008501.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
