Assay Details
Gene Symbol: A930007I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043087.1 | - | 302 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | programmed cell death 1 ligand 2 | Pdcd1lg2 |
B7-DC, Btdc, F730015O22Rik, PD-L2 |
-26.9kb | Anti-sense | Chr. 19 - 29406240 - 29472927 on NCBI Build GRCm38 |
Downstream | RAB6A GEF complex partner 1 | Ric1 |
C030046E11Rik, C130057E09Rik, Kiaa1432 |
1.3kb | Anti-sense | Chr. 19 - 29522282 - 29605921 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
