Assay Details
Gene Symbol: 4930429D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040699.1 | 6 - 7 | 567 | 63 | ||
GenBank mRNA | AK015233.1 | - | 6 - 7 | 568 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitogen-activated protein kinase 10 | Mapk10 |
C230008H04Rik, JNK3, JNK3B1, JNK3B2, SAPK(beta), Serk2, p493F12, p54bSAPK |
-19.0kb | Sense | Chr. 5 - 102907948 - 103211339 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700021F02 gene | 1700021F02Rik | 105.7kb | Anti-sense | Chr. 5 - 103346535 - 103348302 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038147 | AK015233.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824256 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830008 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139921 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630629 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319929 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945274 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135428 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42859 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905894 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127448 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181990 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739757 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455892 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55955 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35260 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99206 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92227 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111222 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038147 | AK015233.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05677929 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111222 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05677970 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05677995 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05677945 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116099 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05677995 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04575983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945739 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23945744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04604463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04802971 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1431990_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431990_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431990_at | - | - |
Mouse Expression Set 430 | 1431990_at | - | - |
Mouse Expression Set 430 | 1431990_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431990_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431990_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 452612 | - | - |
