Assay Details
| Gene Symbol: Gatc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029645.3 | NP_083921.1 | 1 - 2 | 316 | 74 | |
| GenBank mRNA | AK034326.1 | - | 1 - 2 | 316 | 74 | |
| BC021775.1 | - | 1 - 2 | 309 | 4158979 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatc
Genomic Map