Assay Details
Gene Symbol: A330070K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198665.1 | NP_941067.1 | 1 - 2 | 223 | 79 | |
GenBank mRNA | AK039605.1 | - | 1 - 2 | 223 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
