Assay Details
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026555.2 | NP_080831.2 | 4 - 5 | 741 | 85 | |
GenBank mRNA | AK003918.1 | - | 3 - 4 | 411 | 85 | |
AK077943.1 | - | 4 - 5 | 681 | 85 | ||
AK082762.1 | - | 4 - 5 | 671 | 85 | ||
AK086705.1 | - | 4 - 5 | 681 | 85 | ||
AK132143.1 | - | 4 - 5 | 675 | 85 | ||
BC005487.1 | - | 3 - 4 | 578 | 85 | ||
BC025602.1 | - | 4 - 5 | 692 | 5250505 | 85 | |
BC055903.1 | - | 4 - 5 | 688 | 4010685 | 85 | |
BC092069.1 | - | 4 - 5 | 639 | 4215821 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
