Assay Details
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026555.2 | NP_080831.2 | 6 - 7 | 1007 | 67 | |
GenBank mRNA | AK003918.1 | - | 5 - 6 | 677 | 67 | |
AK077943.1 | - | 6 - 7 | 947 | 67 | ||
AK082762.1 | - | 6 - 7 | 937 | 67 | ||
AK086705.1 | - | 6 - 7 | 948 | 67 | ||
AK132143.1 | - | 6 - 7 | 941 | 67 | ||
BC005487.1 | - | 5 - 6 | 844 | 67 | ||
BC025602.1 | - | 6 - 7 | 958 | 5250505 | 67 | |
BC055903.1 | - | 6 - 7 | 954 | 4010685 | 67 | |
BC092069.1 | - | 6 - 7 | 905 | 4215821 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046393 | BC025602.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046393 | BC055903.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046393 | BC005487.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046393 | BC092069.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046393 | AK082762.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902097 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549807 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699947 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944140 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125724 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046393 | BC025602.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046393 | BC055903.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046393 | BC005487.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046393 | BC092069.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046393 | AK082762.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25091117 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06181874 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125724 | - | - |
Whole Mouse Genome G4122A | A_52_P223618 | BC025602.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P223618 | BC055903.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P223618 | BC005487.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P223618 | BC092069.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P223618 | AK082762.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P398683 | BC025602.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P398683 | BC005487.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P398683 | BC092069.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P398683 | AK082762.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P398683 | AV129283.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136172 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116325 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125724 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06605018 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06340077 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06192953 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06340080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06257038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06605018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06102693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06102691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05907131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06027049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05972927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06027048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05972931 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06214168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25091073 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10563080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563081 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563083 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563082 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563084 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563079 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10563078 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424382_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10563077 | - | - |
Mouse Genome 430 2.0 Array | 1424382_at | - | - |
Mouse Genome 430 2.0 Array | 1424382_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52377_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52377_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700026831.mm.1 | NM_026555.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700026831.mm.1 | NM_026555.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700026831.mm.1 | TC0700002988.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700026830.mm.1 | NM_026555.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700026830.mm.1 | NM_026555.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700026830.mm.1 | TC0700002988.mm.1 | MTA TC |
Mouse Expression Set 430 | 1424382_at | - | - |
Mouse Expression Set 430 | 1424382_PM_at | - | - |
Murine Genome U74v2 Set | 96347_PM_at | - | - |
Murine Genome U74v2 Set | 96347_at | - | - |
Mouse Genome 430A 2.0 Array | 1424382_at | - | - |
Mouse Genome 430A 2.0 Array | 1424382_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424382_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424382_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5420400 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5420400 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005420400 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2723718 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2723718 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246312 | - | - |
mouse-6 v1.1 (Illumina) | scl31391.7.1_2-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5420400 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl31391.7.1_2-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420400 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2723718 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723718 | - | - |
