Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 11 - 12 | 1181 | 87 | |
| GenBank mRNA | AK010534.1 | - | 11 - 12 | 1193 | 87 | |
| AK031738.1 | - | 11 - 12 | 1182 | 87 | ||
| AK086646.1 | - | 11 - 12 | 1140 | 87 | ||
| AK155236.1 | - | 11 - 12 | 1179 | 87 | ||
| AK186815.1 | - | 2 - 3 | 118 | 87 | ||
| BC008128.1 | - | 7 - 8 | 724 | 87 | ||
| BC050775.1 | - | 11 - 12 | 1160 | 6534023 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map