Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 12 - 13 | 1374 | 131 | |
| GenBank mRNA | AK010534.1 | - | 12 - 13 | 130 | ||
| AK031738.1 | - | 12 - 13 | 1375 | 131 | ||
| AK086646.1 | - | 12 - 13 | 1333 | 131 | ||
| AK155236.1 | - | 12 - 13 | 1372 | 131 | ||
| BC008128.1 | - | 8 - 9 | 917 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map