Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 13 - 14 | 1456 | 128 | |
| GenBank mRNA | AK010534.1 | - | 13 - 14 | 1467 | 128 | |
| AK031738.1 | - | 13 - 14 | 1457 | 128 | ||
| AK086646.1 | - | 13 - 14 | 1415 | 128 | ||
| AK155236.1 | - | 13 - 14 | 1454 | 128 | ||
| BC008128.1 | - | 9 - 10 | 999 | 128 | ||
| BC050775.1 | - | 13 - 14 | 1447 | 6534023 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map