Assay Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 9 - 10 | 997 | 155 | |
| GenBank mRNA | AK010534.1 | - | 9 - 10 | 1009 | 155 | |
| AK031738.1 | - | 9 - 10 | 998 | 155 | ||
| AK086646.1 | - | 9 - 10 | 956 | 155 | ||
| AK155236.1 | - | 9 - 10 | 995 | 155 | ||
| BC008128.1 | - | 5 - 6 | 540 | 155 | ||
| BC050775.1 | - | 9 - 10 | 976 | 6534023 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt3
Genomic Map