Assay Details
| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008422.1 | NP_001008422.1 | 6 - 7 | 657 | 77 | |
| GenBank mRNA | BC042576.1 | - | 6 - 7 | 515 | 4160122 | 77 |
| BC043138.1 | - | 6 - 7 | 687 | 6821344 | 77 | |
| BC079873.1 | - | 6 - 7 | 675 | 77 | ||
| BC085153.1 | - | 6 - 7 | 657 | 6837651 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf1
Genomic Map