Assay Details
| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008422.1 | NP_001008422.1 | 8 - 9 | 3408 | 71 | |
| GenBank mRNA | BC042576.1 | - | 8 - 9 | 3266 | 4160122 | 71 |
| BC043138.1 | - | 8 - 9 | 3438 | 6821344 | 71 | |
| BC054818.1 | - | 7 - 8 | 5688 | 71 | ||
| BC079873.1 | - | 8 - 9 | 3425 | 71 | ||
| BC085153.1 | - | 8 - 9 | 3408 | 6837651 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf1
Genomic Map