Assay Details
| Gene Symbol: Lipn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027340.2 | NP_081616.1 | 2 - 3 | 345 | 111 | |
| GenBank mRNA | AK008974.1 | - | 2 - 3 | 174 | 111 | |
| AK154333.1 | - | 2 - 3 | 346 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008974.1 transcripts.
Set Membership: |
|

Lipn
Genomic Map