Assay Details
Gene Symbol: Tead2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285498.1 | NP_001272427.1 | 5 - 6 | 596 | 65 | |
NM_001285500.1 | NP_001272429.1 | 5 - 6 | 449 | 65 | ||
NM_011565.2 | NP_035695.1 | 5 - 6 | 554 | 65 | ||
GenBank mRNA | AK132009.1 | - | 4 - 5 | 489 | 65 | |
AK137064.1 | - | 4 - 5 | 1837 | 65 | ||
AK143155.1 | - | 4 - 5 | 1770 | 65 | ||
AK144857.1 | - | 5 - 6 | 516 | 65 | ||
AK146595.1 | - | 5 - 6 | 516 | 65 | ||
BC050217.1 | - | 4 - 5 | 374 | 30021393 | 65 | |
D50563.1 | - | 5 - 6 | 554 | 65 | ||
X94442.1 | - | 4 - 5 | 468 | 65 | ||
Y10026.1 | - | 5 - 6 | 518 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
