Assay Details
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019428.3 | NP_062301.3 | 5 - 6 | 365 | 156 | |
GenBank mRNA | AK004137.1 | - | 5 - 6 | 363 | 156 | |
AK160536.1 | - | 5 - 6 | 292 | 156 | ||
AK169253.1 | - | 5 - 6 | 292 | 156 | ||
BC062146.1 | - | 5 - 6 | 350 | 6518236 | 156 | |
CN722047.1 | - | 5 - 6 | 412 | 156 | ||
U95123.1 | - | 5 - 6 | 344 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
