Assay Details
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019428.3 | NP_062301.3 | 10 - 11 | 715 | 87 | |
GenBank mRNA | AK160536.1 | - | 10 - 11 | 642 | 87 | |
AK169253.1 | - | 10 - 11 | 642 | 87 | ||
BC034329.1 | - | 5 - 6 | 332 | 87 | ||
BC062146.1 | - | 10 - 11 | 700 | 6518236 | 87 | |
U95123.1 | - | 10 - 11 | 694 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131530 | BC034329.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131530 | BC062146.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131530 | U95123.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131530 | AK169253.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131530 | NM_019428.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922454 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368391 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181717 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277671 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114789 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379735 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164006 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309191 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112273 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845899 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558800 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270689 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563486 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575799 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46196 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127002 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129110 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164581 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131530 | BC034329.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131530 | BC062146.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131530 | U95123.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131530 | AK169253.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131530 | NM_019428.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227022 | BC034329.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227022 | BC062146.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227022 | U95123.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227022 | AK169253.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227022 | NM_019428.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P227022 | BC034329.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P227022 | BC062146.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P227022 | U95123.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P227022 | AK169253.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P227022 | NM_019428.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131530 | BC034329.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131530 | BC062146.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131530 | U95123.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131530 | AK169253.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131530 | NM_019428.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122230 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14298405 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14298404 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14371742 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14371741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32088185 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10462674 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462672 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462677 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462676 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462679 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462671 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462681 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462678 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462675 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462680 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10462682 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423373_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447970_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10462670 | - | - |
Mouse Genome 430 2.0 Array | 1447970_at | - | - |
Mouse Genome 430 2.0 Array | 1423373_at | - | - |
Mouse Genome 430 2.0 Array | 1447970_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1423373_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54364_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54364_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900004545.mm.1 | NM_019428.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004545.mm.1 | NM_019428.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004545.mm.1 | TC1900000516.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900004544.mm.1 | NM_019428.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004544.mm.1 | NM_019428.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004544.mm.1 | TC1900000516.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900004546.mm.1 | NM_019428.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004546.mm.1 | NM_019428.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900004546.mm.1 | TC1900000516.mm.1 | MTA TC |
Mouse Expression Set 430 | 1447970_at | - | - |
Mouse Expression Set 430 | 1423373_at | - | - |
Mouse Expression Set 430 | 1447970_PM_at | - | - |
Mouse Expression Set 430 | 1423373_PM_at | - | - |
Murine Genome U74v2 Set | 102111_f_at | - | - |
Murine Genome U74v2 Set | 102111_PM_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1423373_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1447970_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423373_at | - | - |
Mouse Genome 430A 2.0 Array | 1447970_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423373_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1447970_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1447970_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423373_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3870504 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003870504 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232428 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1232428 | - | - |
mouse-6 v1.1 (Illumina) | scl53186.10.89_4-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3870504 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl53186.10.89_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870504 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1232428 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3007195 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3007195 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232428 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53186.10.89_4-S | - | - |
