Assay Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146133.1 | - | 9 - 10 | 1053 | 67 | |
| BC145590.1 | - | 9 - 10 | 944 | 9054156 | 67 | |
| BC145591.1 | - | 9 - 10 | 941 | 9054157 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf5
Genomic Map