Assay Details
| Gene Symbol: Gjc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080450.4 | NP_536698.2 | 2 - 3 | 880 | 68 | |
| GenBank mRNA | AK034037.1 | - | 2 - 3 | 845 | 68 | |
| AK045577.1 | - | 2 - 3 | 1020 | 68 | ||
| AK047368.1 | - | 2 - 3 | 881 | 68 | ||
| AK147913.1 | - | 2 - 3 | 1020 | 68 | ||
| BC125305.1 | - | 2 - 3 | 867 | 40129820 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108948 | AK147913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2108948 | NM_080450.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 685397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19923 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108109 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108948 | AK147913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2108948 | NM_080450.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413721 | AY289210.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413721 | AY293564.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413721 | AY297108.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413721 | BC125305.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413721 | AK147913.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128386 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05761955 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108109 | - | - |
| Whole Mouse Genome G4122A | A_51_P413721 | AY289210.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413721 | AY293564.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413721 | AY297108.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413721 | BC125305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413721 | AK147913.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105512 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108109 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04946885 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04817545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04946878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04816405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04946877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04946877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04537420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04537419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04486497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04537417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04209967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04174368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04702396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04481287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04816405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04722449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04439715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04473931 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534963 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422042_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534960 | - | - |
| Mouse Genome 430 2.0 Array | 1422042_at | - | - |
| Mouse Genome 430 2.0 Array | 1422042_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445523_at | - | - |
| Mouse Genome 430 2.0 Array | 1445523_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 118446_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 118446_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031610.mm.1 | TC0500003468.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031610.mm.1 | NM_080450.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031610.mm.1 | NM_080450.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031609.mm.1 | TC0500003468.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031609.mm.1 | NM_080450.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031609.mm.1 | NM_080450.4 | Refseq |
| Mouse Expression Set 430 | 1422042_at | - | - |
| Mouse Expression Set 430 | 1422042_PM_at | - | - |
| Mouse Expression Set 430 | 1445523_at | - | - |
| Mouse Expression Set 430 | 1445523_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422042_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422042_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422042_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422042_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110022 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685506 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685506 | - | - |
| mouse-6 v1.1 (Illumina) | scl25863.3_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25863.3_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110022 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685506 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685506 | - | - |

Gjc3
Genomic Map