Assay Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053262.3 | NP_444492.1 | 3 - 4 | 505 | 66 | |
| GenBank mRNA | AF304306.1 | - | 3 - 4 | 501 | 66 | |
| AK049355.1 | - | 3 - 4 | 549 | 66 | ||
| AK154803.1 | - | 3 - 4 | 520 | 66 | ||
| AK155174.1 | - | 3 - 4 | 520 | 66 | ||
| AK155202.1 | - | 3 - 4 | 525 | 66 | ||
| AK155327.1 | - | 3 - 4 | 525 | 66 | ||
| AK158327.1 | - | 3 - 4 | 520 | 66 | ||
| AK170939.1 | - | 3 - 4 | 520 | 66 | ||
| AY053570.1 | - | 3 - 4 | 519 | 66 | ||
| BC038340.1 | - | 3 - 4 | 505 | 4458725 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b11
Genomic Map