Assay Details
| Gene Symbol: Hsd17b11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053262.3 | NP_444492.1 | 4 - 5 | 618 | 74 | |
| GenBank mRNA | AF304306.1 | - | 4 - 5 | 614 | 74 | |
| AK049355.1 | - | 4 - 5 | 662 | 74 | ||
| AK154803.1 | - | 4 - 5 | 633 | 74 | ||
| AK155174.1 | - | 4 - 5 | 633 | 74 | ||
| AK155202.1 | - | 4 - 5 | 638 | 74 | ||
| AK155327.1 | - | 4 - 5 | 638 | 74 | ||
| AK158327.1 | - | 4 - 5 | 633 | 74 | ||
| AK170939.1 | - | 4 - 5 | 633 | 74 | ||
| AY053570.1 | - | 4 - 5 | 632 | 74 | ||
| BC038340.1 | - | 4 - 5 | 618 | 4458725 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b11
Genomic Map