Assay Details
| Gene Symbol: Sorbs2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045739.1 | 2 - 3 | 271 | 63 | ||
| GenBank mRNA | AK084672.1 | - | 2 - 3 | 272 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30759 |
Gm30759 | -181.3kb | Sense | Chr. 8 - 45526202 - 45560857 on NCBI Build GRCm38 | |
| Downstream | PDZ and LIM domain 3 | Pdlim3 |
AI463105, ALP, Actn2lp |
139.6kb | Anti-sense | Chr. 8 - 45885453 - 45919548 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Sorbs2os
Genomic Map