Assay Details
| Gene Symbol: Gna14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008137.4 | NP_032163.3 | 4 - 5 | 1622 | 76 | |
| GenBank mRNA | AK035320.1 | - | 3 - 4 | 462 | 76 | |
| AK085444.1 | - | 4 - 5 | 1528 | 76 | ||
| AK160808.1 | - | 4 - 5 | 1216 | 76 | ||
| AK168498.1 | - | 4 - 5 | 1278 | 76 | ||
| BC027015.1 | - | 4 - 5 | 1090 | 5375817 | 76 | |
| M80631.1 | - | 4 - 5 | 751 | 76 | ||
More Information
Set Membership: |
|

Gna14
Genomic Map