Assay Details
| Gene Symbol: Pip5k1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008846.2 | NP_032872.1 | 3 - 4 | 220 | 77 | |
| GenBank mRNA | AK077334.1 | - | 3 - 4 | 231 | 77 | |
| AK133809.1 | - | 3 - 4 | 215 | 77 | ||
| AK146100.1 | - | 3 - 4 | 220 | 77 | ||
| AK149354.1 | - | 3 - 4 | 220 | 77 | ||
| AK164552.1 | - | 3 - 4 | 203 | 77 | ||
| BC034864.1 | - | 3 - 4 | 234 | 3326897 | 77 | |
| D86176.1 | - | 2 - 3 | 153 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1b
Genomic Map