Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500027242.mm.1 |
TC0500003060.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027242.mm.1 |
NM_001159941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027242.mm.1 |
NM_001159941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027242.mm.1 |
NM_026145.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027242.mm.1 |
NM_026145.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027243.mm.1 |
TC0500003060.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500027243.mm.1 |
NM_001159941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027243.mm.1 |
NM_001159941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027243.mm.1 |
NM_026145.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500027243.mm.1 |
NM_026145.4 |
Refseq |