Assay Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007651.3 | NP_031677.1 | 4 - 5 | 532 | 102 | |
| GenBank mRNA | AA177409.1 | - | 4 - 5 | 420 | 102 | |
| AK156510.1 | - | 4 - 5 | 439 | 102 | ||
| BC021310.1 | - | 4 - 5 | 446 | 5010267 | 102 | |
| BC052905.1 | - | 4 - 5 | 397 | 30078163 | 102 | |
| X97227.1 | - | 3 - 4 | 333 | 102 | ||
| Z16071.1 | - | 3 - 4 | 318 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd53
Genomic Map