Assay Details
| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286396.1 | NP_001273325.1 | 1 - 2 | 44 | 74 | |
| GenBank mRNA | BC018377.1 | - | 1 - 2 | 44 | 4242375 | 74 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC018377.1, NM_001286396.1 transcripts.
Set Membership: |
|

Dao
Genomic Map