Assay Details
Gene Symbol: Prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044382.1 | NP_001037847.1 | 2 - 3 | 255 | 90 | |
GenBank mRNA | AF487281.1 | - | 1 - 2 | 125 | 90 | |
AK044344.1 | - | 1 - 2 | 174 | 90 | ||
BC042707.1 | - | 2 - 3 | 170 | 4975079 | 90 | |
BC117006.1 | - | 2 - 3 | 255 | 40126325 | 90 | |
BC117008.1 | - | 2 - 3 | 255 | 40126327 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P306357 | BC042707.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P306357 | AK044344.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P306357 | BC042707.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P306357 | AK044344.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163183 | BC117006.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163183 | BC117008.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163183 | NM_001044382.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124057 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173637 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953136 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164226 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163183 | BC117006.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163183 | BC117008.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163183 | NM_001044382.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164226 | - | - |
Whole Mouse Genome G4122A | A_52_P306357 | BC042707.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P306357 | AK044344.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P306357 | BC042707.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P306357 | AK044344.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P482724 | BC042707.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P482724 | AF487281.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P482724 | AK044344.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122413 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22405128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02601578 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02641031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02641055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02601582 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405113 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405052 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02892446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02908362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02641050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02402229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02908363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02641048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405102 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405074 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405103 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02402232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02641040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02933182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03014636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02670662 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405100 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405131 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02908364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405056 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02581875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22405091 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10501088 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501090 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501086 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501089 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501087 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10501085 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10501084 | - | - |
Affymetrix Mouse Clariom S Array | PSR0300021045.mm.1 | NM_001044382.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021045.mm.1 | NM_001044382.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021045.mm.1 | TC0300002702.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300021041.mm.1 | NM_001044382.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021041.mm.1 | NM_001044382.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300021041.mm.1 | TC0300002702.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300021040.mm.1 | TC0300002702.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 323066 | - | - |
ABI Mouse Genome Survey Microarray | 701345 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_2886025 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2886025 | - | - |
