Assay Details
Gene Symbol: Cyp11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033229.3 | NP_001028401.2 | 4 - 5 | 837 | 87 | |
GenBank mRNA | AK142439.1 | - | 4 - 5 | 814 | 87 | |
AK163083.1 | - | 4 - 5 | 813 | 87 | ||
AK168839.1 | - | 4 - 5 | 814 | 87 | ||
BC139248.1 | - | 4 - 5 | 837 | 8862270 | 87 | |
BC139250.1 | - | 4 - 5 | 837 | 8862272 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
