Assay Details
Gene Symbol: Cyp11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033229.3 | NP_001028401.2 | 7 - 8 | 1232 | 75 | |
GenBank mRNA | AK142439.1 | - | 7 - 8 | 1209 | 75 | |
AK163083.1 | - | 7 - 8 | 1208 | 75 | ||
AK168839.1 | - | 7 - 8 | 1209 | 75 | ||
BC139248.1 | - | 7 - 8 | 1232 | 8862270 | 75 | |
BC139250.1 | - | 7 - 8 | 1232 | 8862272 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
regulation of blood volume by renal aldosterone
C21-steroid hormone biosynthetic process
glucocorticoid biosynthetic process
aldosterone biosynthetic process
potassium ion homeostasis
sodium ion homeostasis
oxidation-reduction process
steroid 11-beta-monooxygenase activity
heme binding
corticosterone 18-monooxygenase activity
regulation of blood volume by renal aldosterone
C21-steroid hormone biosynthetic process
glucocorticoid biosynthetic process
aldosterone biosynthetic process
potassium ion homeostasis
sodium ion homeostasis
oxidation-reduction process
steroid 11-beta-monooxygenase activity
heme binding
corticosterone 18-monooxygenase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156963 | AK163083.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156963 | NM_001033229.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156963 | AK142439.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107392 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816544 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434380 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156963 | AK163083.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156963 | NM_001033229.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156963 | AK142439.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08418458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339884 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12017062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339922 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12296583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339941 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339912 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12017067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11931230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12017089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339920 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12017093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339930 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11931226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339979 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339971 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11936357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12296586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11936355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11936352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12017087 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12311313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339918 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339907 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10429542 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429545 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429544 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429543 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429541 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429546 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456362_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10429538 | - | - |
Mouse Genome 430 2.0 Array | 1456362_at | - | - |
Mouse Genome 430 2.0 Array | 1456362_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110115_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110115_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500012995.mm.1 | NM_001033229.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012995.mm.1 | NM_001033229.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012995.mm.1 | TC1500001723.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012993.mm.1 | NM_001033229.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012993.mm.1 | NM_001033229.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012993.mm.1 | TC1500001723.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012994.mm.1 | NM_001033229.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012994.mm.1 | NM_001033229.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012994.mm.1 | TC1500001723.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012992.mm.1 | NM_001033229.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012992.mm.1 | NM_001033229.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012992.mm.1 | TC1500001723.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456362_at | - | - |
Mouse Expression Set 430 | 1456362_PM_at | - | - |
Murine Genome U74v2 Set | 166619_f_at | - | - |
Murine Genome U74v2 Set | 166619_PM_f_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456362_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456362_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 598701 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_2993882 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2993882 | - | - |
